siRNA Details
| Gene Symbol: Wdsub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014179.1 | NP_001014201.1 | 2 | 358 | ||
| XM_006234208.2 | XP_006234270.1 | 2 | 618 | |||
| XM_006234209.2 | XP_006234271.1 | 2 | 617 | |||
| XM_006234211.2 | XP_006234273.1 | 2 | 339 | |||
| GenBank mRNA | BC089856.1 | - | 2 | 358 | 7381399 | |

Wdsub1
Genomic Map