siRNA Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012184.1 | NP_001012184.1 | 10 | 1355 | ||
| XM_006234234.2 | XP_006234296.1 | 11 | 1160 | |||
| XM_006234235.2 | XP_006234297.1 | 11 | 1453 | |||
| XM_006234236.2 | XP_006234298.1 | 11 | 1252 | |||
| XM_006234237.2 | XP_006234299.1 | 11 | 2012 | |||
| XM_006234238.2 | XP_006234300.1 | 10 | 1112 | |||
| XM_006234239.2 | XP_006234301.1 | 12 | 1650 | |||
| XM_006234241.2 | XP_006234303.1 | 10 | 1202 | |||
| XM_008761860.1 | XP_008760082.1 | 11 | 1741 | |||
| GenBank mRNA | BC087094.1 | - | 10 | 1355 | 7191334 | |

Rbms1
Genomic Map