siRNA Details
| Gene Symbol: Bbs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108583.2 | NP_001102053.1 | 5, 6 | 426 | ||
| GenBank mRNA | BC167024.1 | - | 5, 6 | 426 | 7123978 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs5| Gene Symbol: Bbs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108583.2 | NP_001102053.1 | 5, 6 | 426 | ||
| GenBank mRNA | BC167024.1 | - | 5, 6 | 426 | 7123978 | |