siRNA Details
| Gene Symbol: Cir1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007799.1 | NP_001007800.1 | 5, 6 | 320 | ||
| GenBank mRNA | BC085869.1 | - | 5, 6 | 390 | 7191148 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cir1| Gene Symbol: Cir1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007799.1 | NP_001007800.1 | 5, 6 | 320 | ||
| GenBank mRNA | BC085869.1 | - | 5, 6 | 390 | 7191148 | |