siRNA Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077429.1 | NP_001070897.1 | 10, 11 | 922 | ||
| XM_006234377.2 | XP_006234439.1 | 10, 11 | 1004 | |||
| XM_006234378.2 | XP_006234440.1 | 11, 12 | 1025 | |||
| XM_006234379.2 | XP_006234441.1 | 10, 11 | 1052 | |||
| XM_006234380.2 | XP_006234442.1 | 10, 11 | 1124 | |||
| XM_006234381.2 | XP_006234443.1 | 10, 11 | 1217 | |||
| XM_006234382.2 | XP_006234444.1 | 11, 12 | 1145 | |||
| XM_006234385.2 | XP_006234447.1 | 10, 11 | 1006 | |||
| XM_008761909.1 | XP_008760131.1 | 10, 11 | 2101 | |||
| XM_008761910.1 | XP_008760132.1 | 10, 11 | 1006 | |||
| GenBank mRNA | BC126100.1 | - | 10, 11 | 922 | 7132654 | |
| FQ218047.1 | - | 9, 10 | 801 | |||

Lnp
Genomic Map