siRNA Details
Gene Symbol: Zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010963.1 | NP_001010963.1 | 3 | 312 | ||
GenBank mRNA | AY377983.1 | - | 3 | 268 | ||
BC088438.1 | - | 3 | 312 | 7126102 | ||
FQ232115.1 | - | 3 | 391 | |||
FQ233565.1 | - | 3 | 414 |
