siRNA Details
Gene Symbol: Zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010963.1 | NP_001010963.1 | 9 | 1119 | ||
GenBank mRNA | AY377983.1 | - | 9 | 1075 | ||
BC088438.1 | - | 9 | 1119 | 7126102 | ||
FQ212631.1 | - | 4 | 407 |
