siRNA Details
| Gene Symbol: Creb3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005562.1 | NP_001005562.1 | 6 | 1285 | ||
| XM_006234584.1 | XP_006234646.1 | 6 | 1341 | |||
| GenBank mRNA | BC081951.1 | - | 6 | 1285 | 7122657 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Creb3l1| Gene Symbol: Creb3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005562.1 | NP_001005562.1 | 6 | 1285 | ||
| XM_006234584.1 | XP_006234646.1 | 6 | 1341 | |||
| GenBank mRNA | BC081951.1 | - | 6 | 1285 | 7122657 | |