siRNA Details
| Gene Symbol: Creb3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005562.1 | NP_001005562.1 | 7, 8 | 1364 | ||
| XM_006234584.1 | XP_006234646.1 | 7, 8 | 1420 | |||
| GenBank mRNA | BC081951.1 | - | 7, 8 | 1364 | 7122657 | |

Creb3l1
Genomic Map