siRNA Details
| Gene Symbol: Alkbh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014180.1 | NP_001014202.1 | 9 | 814 | ||
| XM_006234585.2 | XP_006234647.1 | 9 | 892 | |||
| GenBank mRNA | BC083756.1 | - | 9 | 814 | 7192721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkbh3| Gene Symbol: Alkbh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014180.1 | NP_001014202.1 | 9 | 814 | ||
| XM_006234585.2 | XP_006234647.1 | 9 | 892 | |||
| GenBank mRNA | BC083756.1 | - | 9 | 814 | 7192721 | |