siRNA Details
| Gene Symbol: Prr5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080150.1 | NP_001073619.1 | 4 | 630 | ||
| XM_006234647.2 | XP_006234709.1 | 4 | 1596 | |||
| XM_006234648.2 | XP_006234710.1 | 4 | 897 | |||
| GenBank mRNA | BC129104.1 | - | 4 | 630 | 7100178 | |

Prr5l
Genomic Map