siRNA Details
| Gene Symbol: Prr5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080150.1 | NP_001073619.1 | 6 | 784 | ||
| XM_006234647.2 | XP_006234709.1 | 6 | 1750 | |||
| XM_006234648.2 | XP_006234710.1 | 6 | 1051 | |||
| GenBank mRNA | AY724495.1 | - | 1 | 58 | ||
| BC129104.1 | - | 6 | 784 | 7100178 | ||

Prr5l
Genomic Map