siRNA Details
| Gene Symbol: Lmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037358.2 | NP_001032435.2 | 4 | 735 | ||
| NM_001244779.1 | NP_001231708.1 | 4 | 625 | |||
| NM_001244780.1 | NP_001231709.1 | 4 | 600 | |||
| NM_001244781.1 | NP_001231710.1 | 4 | 630 | |||
| GenBank mRNA | BC105772.1 | - | 4 | 735 | 7930561 | |
| CK476274.1 | - | 4 | 586 | |||
| CO559731.1 | - | 600 | ||||
| DY319476.1 | - | 4 | 626 | |||

Lmo2
Genomic Map