siRNA Details
| Gene Symbol: Rcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108586.1 | NP_001102056.1 | 5 | 1192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcn1| Gene Symbol: Rcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108586.1 | NP_001102056.1 | 5 | 1192 | ||