siRNA Details
| Gene Symbol: Ccdc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108587.1 | NP_001102057.1 | 5 | 1580 | ||
| GenBank mRNA | FQ225525.1 | - | 6 | 951 | ||
| Gene Symbol: LOC102551066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234694.1 | XP_006234756.1 | 2 | 234 | ||

Ccdc34
Genomic Map