siRNA Details
| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014181.1 | NP_001014203.1 | 529 | |||
| GenBank mRNA | BC087148.1 | - | 529 | 7131040 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dph6| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014181.1 | NP_001014203.1 | 529 | |||
| GenBank mRNA | BC087148.1 | - | 529 | 7131040 | ||