siRNA Details
| Gene Symbol: Chac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173437.1 | NP_001166908.1 | 3 | 562 | ||
| GenBank mRNA | EF688602.1 | - | 3 | 562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chac1| Gene Symbol: Chac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173437.1 | NP_001166908.1 | 3 | 562 | ||
| GenBank mRNA | EF688602.1 | - | 3 | 562 | ||