siRNA Details
| Gene Symbol: Vps39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012186.2 | NP_001012186.2 | 18 | 1966 | ||
| XM_006234782.2 | XP_006234844.1 | 19 | 2010 | |||
| GenBank mRNA | BC087048.1 | - | 17 | 2387 | 7112790 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps39| Gene Symbol: Vps39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012186.2 | NP_001012186.2 | 18 | 1966 | ||
| XM_006234782.2 | XP_006234844.1 | 19 | 2010 | |||
| GenBank mRNA | BC087048.1 | - | 17 | 2387 | 7112790 | |