siRNA Details
| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013204.1 | NP_001013222.1 | 8 | 705 | ||
| XM_006234859.1 | XP_006234921.1 | 8 | 800 | |||
| GenBank mRNA | BC091256.1 | - | 8 | 705 | 7375652 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccndbp1| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013204.1 | NP_001013222.1 | 8 | 705 | ||
| XM_006234859.1 | XP_006234921.1 | 8 | 800 | |||
| GenBank mRNA | BC091256.1 | - | 8 | 705 | 7375652 | |