siRNA Details
| Gene Symbol: Mall | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014182.1 | NP_001014204.1 | 3 | 410 | ||
| XM_008762181.1 | XP_008760403.1 | 3 | 447 | |||
| GenBank mRNA | BC090079.1 | - | 3 | 410 | 7126923 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mall| Gene Symbol: Mall | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014182.1 | NP_001014204.1 | 3 | 410 | ||
| XM_008762181.1 | XP_008760403.1 | 3 | 447 | |||
| GenBank mRNA | BC090079.1 | - | 3 | 410 | 7126923 | |