siRNA Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191854.1 | NP_001178783.1 | 12 | 1179 | ||
| XM_006234976.2 | XP_006235038.1 | 12 | 1383 | |||
| XM_008762182.1 | XP_008760404.1 | 12 | 1381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem87b| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191854.1 | NP_001178783.1 | 12 | 1179 | ||
| XM_006234976.2 | XP_006235038.1 | 12 | 1383 | |||
| XM_008762182.1 | XP_008760404.1 | 12 | 1381 | |||