siRNA Details
Gene Symbol: Lamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014183.1 | NP_001014205.1 | 6 | 1061 | ||
GenBank mRNA | BC087680.1 | - | 6 | 1061 | 7318802 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014183.1 | NP_001014205.1 | 6 | 1061 | ||
GenBank mRNA | BC087680.1 | - | 6 | 1061 | 7318802 |