siRNA Details
| Gene Symbol: Snrpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108592.2 | NP_001102062.1 | 6 | 517 | ||
| XM_006235116.2 | XP_006235178.1 | 6 | 678 | |||
| GenBank mRNA | BC168760.1 | - | 6 | 517 | 6888886 | |
| FQ213679.1 | - | 6 | 549 | |||

Snrpb2
Genomic Map