siRNA Details
| Gene Symbol: Syndig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025020.1 | NP_001020191.1 | 3 | 513 | ||
| GenBank mRNA | BC092131.1 | - | 3 | 513 | 7312153 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syndig1| Gene Symbol: Syndig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025020.1 | NP_001020191.1 | 3 | 513 | ||
| GenBank mRNA | BC092131.1 | - | 3 | 513 | 7312153 | |