siRNA Details
| Gene Symbol: Dusp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108598.2 | NP_001102068.2 | 3, 4 | 352 | ||
| NM_001244784.1 | NP_001231713.1 | 3, 4 | 352 | |||
| GenBank mRNA | BC168211.1 | - | 3, 4 | 181 | 7135378 | |
| CB576376.1 | - | 3, 4 | 229 | |||
| CB585518.1 | - | 3, 4 | 352 | |||
| CB736780.1 | - | 3, 4 | 431 | |||

Dusp15
Genomic Map