siRNA Details
| Gene Symbol: Snta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100901.1 | NP_001094371.1 | 3 | 614 | ||
| XM_006235326.2 | XP_006235388.1 | 3 | 653 | |||
| GenBank mRNA | BC169101.1 | - | 3 | 614 | 5599966 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snta1| Gene Symbol: Snta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100901.1 | NP_001094371.1 | 3 | 614 | ||
| XM_006235326.2 | XP_006235388.1 | 3 | 653 | |||
| GenBank mRNA | BC169101.1 | - | 3 | 614 | 5599966 | |