siRNA Details
| Gene Symbol: Snta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100901.1 | NP_001094371.1 | 4 | 896 | ||
| XM_006235326.2 | XP_006235388.1 | 4 | 935 | |||
| GenBank mRNA | BC100086.1 | - | 2 | 192 | ||
| BC169101.1 | - | 4 | 896 | 5599966 | ||

Snta1
Genomic Map