siRNA Details
| Gene Symbol: Map1lc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199500.2 | NP_955794.1 | 4 | 486 | ||
| GenBank mRNA | AY206668.1 | - | 4 | 434 | ||
| BC086389.1 | - | 4 | 447 | 7302206 | ||
| FM036735.1 | - | 3 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map1lc3a| Gene Symbol: Map1lc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199500.2 | NP_955794.1 | 4 | 486 | ||
| GenBank mRNA | AY206668.1 | - | 4 | 434 | ||
| BC086389.1 | - | 4 | 447 | 7302206 | ||
| FM036735.1 | - | 3 | 271 | |||