siRNA Details
| Gene Symbol: Trpc4ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100748.1 | NP_001094218.1 | 10, 11 | 1393 | ||
| GenBank mRNA | AY724540.1 | - | 10, 11 | 1365 | ||
| BC087662.1 | - | 10, 11 | 1346 | |||
| BC166758.1 | - | 10, 11 | 1366 | 7930596 | ||

Trpc4ap
Genomic Map