siRNA Details
| Gene Symbol: Procr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025733.2 | NP_001020904.1 | 4 | 756 | ||
| GenBank mRNA | BC097380.1 | - | 4 | 727 | 7445753 | |
| BC099115.1 | - | 4 | 756 | 7462042 | ||
| FQ220150.1 | - | 4 | 795 | |||
| FQ221148.1 | - | 4 | 764 | |||
| FQ228484.1 | - | 4 | 761 | |||
| FQ228710.1 | - | 4 | 763 | |||
| FQ229445.1 | - | 4 | 793 | |||

Procr
Genomic Map