siRNA Details
Gene Symbol: Snhg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258011.1 | NP_001244940.1 | 2, 3 | 445 | ||
GenBank mRNA | BC094214.1 | - | 2, 3 | 445 | 7314456 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snhg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258011.1 | NP_001244940.1 | 2, 3 | 445 | ||
GenBank mRNA | BC094214.1 | - | 2, 3 | 445 | 7314456 |