siRNA Details
| Gene Symbol: Dhx35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108601.1 | NP_001102071.1 | 9 | 812 | ||
| XM_006235423.2 | XP_006235485.1 | 9 | 818 | |||
| XM_008762359.1 | XP_008760581.1 | 9 | 819 | |||
| XM_008762360.1 | XP_008760582.1 | 5 | 318 | |||

Dhx35
Genomic Map