siRNA Details
| Gene Symbol: Lpin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014184.1 | NP_001014206.1 | 10, 11 | 1524 | ||
| XM_006235487.2 | XP_006235549.1 | 12, 13 | 2949 | |||
| XM_006235489.2 | XP_006235551.1 | 12, 13 | 2300 | |||
| XM_006235490.2 | XP_006235552.1 | 13, 14 | 3094 | |||
| XM_006235491.2 | XP_006235553.1 | 12, 13 | 2560 | |||
| XM_006235493.2 | XP_006235555.1 | 11, 12 | 2094 | |||
| XM_006235494.2 | XP_006235556.1 | 11, 12 | 2743 | |||
| GenBank mRNA | BC089878.1 | - | 10, 11 | 1524 | 7326188 | |

Lpin3
Genomic Map