siRNA Details
| Gene Symbol: Lpin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014184.1 | NP_001014206.1 | 17, 18 | 2312 | ||
| XM_006235487.2 | XP_006235549.1 | 19, 20 | 3737 | |||
| XM_006235489.2 | XP_006235551.1 | 19, 20 | 3088 | |||
| XM_006235490.2 | XP_006235552.1 | 20, 21 | 3882 | |||
| XM_006235491.2 | XP_006235553.1 | 19, 20 | 3348 | |||
| XM_006235493.2 | XP_006235555.1 | 18, 19 | 2882 | |||
| XM_006235494.2 | XP_006235556.1 | 18, 19 | 3531 | |||
| GenBank mRNA | BC089878.1 | - | 17, 18 | 2312 | 7326188 | |

Lpin3
Genomic Map