siRNA Details
| Gene Symbol: Emilin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109901.2 | NP_001103371.1 | 3 | 461 | ||
| XM_006235495.1 | XP_006235557.1 | 3 | 1135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emilin3| Gene Symbol: Emilin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109901.2 | NP_001103371.1 | 3 | 461 | ||
| XM_006235495.1 | XP_006235557.1 | 3 | 1135 | |||