siRNA Details
| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014185.2 | NP_001014207.2 | 4 | 598 | ||
| GenBank mRNA | BC087121.1 | - | 4 | 589 | 7106749 | |
| FM060375.1 | - | 4 | 598 | |||
| FM081417.1 | - | 2 | 122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srsf6| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014185.2 | NP_001014207.2 | 4 | 598 | ||
| GenBank mRNA | BC087121.1 | - | 4 | 589 | 7106749 | |
| FM060375.1 | - | 4 | 598 | |||
| FM081417.1 | - | 2 | 122 | |||