siRNA Details
| Gene Symbol: Svs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102403.1 | NP_001095873.1 | 3 | 273 | ||
| Gene Symbol: LOC103694891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773872.1 | XP_008772094.1 | 3 | 251 | ||

Svs6
Genomic Map