siRNA Details
| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108604.1 | NP_001102074.1 | 8, 9 | 1015 | ||
| XM_006235598.2 | XP_006235660.1 | 7, 8 | 957 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbpjl| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108604.1 | NP_001102074.1 | 8, 9 | 1015 | ||
| XM_006235598.2 | XP_006235660.1 | 7, 8 | 957 | |||