siRNA Details
| Gene Symbol: HCAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006018.2 | NP_006009.2 | 1 | 405 | ||
| GenBank mRNA | AK290930.1 | - | 1 | 286 | ||
| AK313212.1 | - | 1 | 304 | |||
| BC038955.1 | - | 1 | 295 | |||
| BC047891.1 | - | 1 | 310 | 5750043 | ||
| BI908162.1 | - | 1 | 298 | |||
| D10923.1 | - | 1 | 310 | |||
| DA953971.1 | - | 1 | 324 | |||

HCAR3
Genomic Map