siRNA Details
| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108607.1 | NP_001102077.1 | 19 | 2081 | ||
| XM_006235648.2 | XP_006235710.1 | 20 | 2179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cse1l| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108607.1 | NP_001102077.1 | 19 | 2081 | ||
| XM_006235648.2 | XP_006235710.1 | 20 | 2179 | |||