siRNA Details
| Gene Symbol: Rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001517.1 | NP_001001517.1 | 1 | 125 | ||
| GenBank mRNA | AY603473.1 | - | 1 | 125 | ||
| BC098633.1 | - | 1 | 116 | 7369070 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf114| Gene Symbol: Rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001517.1 | NP_001001517.1 | 1 | 125 | ||
| GenBank mRNA | AY603473.1 | - | 1 | 125 | ||
| BC098633.1 | - | 1 | 116 | 7369070 | ||