siRNA Details
| Gene Symbol: Tfap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201420.2 | NP_958823.2 | 7 | 1363 | ||
| XM_006235691.2 | XP_006235753.1 | 7 | 1332 | |||
| GenBank mRNA | AY510602.1 | - | 7 | 1154 | ||
| BC097260.1 | - | 7 | 1363 | 7443035 | ||

Tfap2c
Genomic Map