siRNA Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033708.1 | NP_001028880.1 | 9 | 767 | ||
| XM_006235692.2 | XP_006235754.1 | 10 | 1078 | |||
| XM_008762517.1 | XP_008760739.1 | 8 | 1369 | |||
| XM_008762518.1 | XP_008760740.1 | 9 | 1961 | |||
| XM_008762519.1 | XP_008760741.1 | 9 | 691 | |||
| GenBank mRNA | BC104721.1 | - | 9 | 767 | 7383476 | |
| BC105758.1 | - | 9 | 774 | 5623260 | ||

Rae1
Genomic Map