siRNA Details
| Gene Symbol: Phactr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214459.2 | NP_999624.1 | 10, 11 | 1481 | ||
| GenBank mRNA | AY500157.1 | - | 10, 11 | 1481 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001007154, NM_001177791, NM_028806, NM_001177790, NM_001177789, and others.

Phactr3
Genomic Map