siRNA Details
| Gene Symbol: Rtel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191857.1 | NP_001178786.1 | 32 | 3567 | ||
| XM_006235772.2 | XP_006235834.1 | 32 | 3614 | |||
| XM_006235773.2 | XP_006235835.1 | 32 | 4023 | |||
| XM_006235774.2 | XP_006235836.1 | 32 | 3614 | |||
| XM_006235775.2 | XP_006235837.1 | 32 | 3575 | |||
| XM_006235776.2 | XP_006235838.1 | 31 | 3497 | |||
| XM_006235778.2 | XP_006235840.1 | 25 | 3153 | |||
| GenBank mRNA | BC086436.1 | - | 5 | 716 | ||

Rtel1
Genomic Map