siRNA Details
| Gene Symbol: Lime1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108614.1 | NP_001102084.1 | 3 | 1135 | ||
| XM_008762521.1 | XP_008760743.1 | 4 | 711 | |||
| GenBank mRNA | BC158665.1 | - | 3 | 625 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lime1| Gene Symbol: Lime1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108614.1 | NP_001102084.1 | 3 | 1135 | ||
| XM_008762521.1 | XP_008760743.1 | 4 | 711 | |||
| GenBank mRNA | BC158665.1 | - | 3 | 625 | ||