siRNA Details
| Gene Symbol: Rbm33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191859.1 | NP_001178788.1 | 801 | |||
| XM_006235845.2 | XP_006235907.1 | 2949 | ||||
| XM_008762624.1 | XP_008760846.1 | 3069 | ||||
| XM_008762625.1 | XP_008760847.1 | 3069 | ||||
| XM_008762626.1 | XP_008760848.1 | 2949 | ||||

Rbm33
Genomic Map