siRNA Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080153.1 | NP_001073622.1 | 3 | 594 | ||
| NM_198781.2 | NP_942076.2 | 3 | 504 | |||
| XM_006235944.2 | XP_006236006.1 | 3 | 1240 | |||
| XM_006235945.2 | XP_006236007.1 | 3 | 631 | |||
| XM_008762651.1 | XP_008760873.1 | 3 | 506 | |||
| GenBank mRNA | AY603378.1 | - | 3 | 594 | ||
| DY313123.1 | - | 3 | 504 | |||

Tmub1
Genomic Map