siRNA Details
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014186.1 | NP_001014208.1 | 10 | 992 | ||
| XM_006235947.2 | XP_006236009.1 | 10 | 1048 | |||
| GenBank mRNA | BC088447.1 | - | 10 | 992 | 7126516 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orc5| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014186.1 | NP_001014208.1 | 10 | 992 | ||
| XM_006235947.2 | XP_006236009.1 | 10 | 1048 | |||
| GenBank mRNA | BC088447.1 | - | 10 | 992 | 7126516 | |