siRNA Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108618.1 | NP_001102088.1 | 3 | 532 | ||
| GenBank mRNA | BC168923.1 | - | 3 | 539 | 7105532 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krit1| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108618.1 | NP_001102088.1 | 3 | 532 | ||
| GenBank mRNA | BC168923.1 | - | 3 | 539 | 7105532 | |